3-(Methylthio)-2-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

81081-500MG 81081-1G 81081-5G
77340-272EA 869.32 USD
77340-272 77358-800 77382-716
3-(Methylthio)-2-propoxybenzoic acid ≥97%
3-(Methylthio)-2-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃S
MW: 226.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768588
CAS Number: 2901066-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR