3-(Methylthio)-2-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

81079-500MG 81079-1G
77340-432EA 1053.69 USD
77340-432 77358-798
3-(Methylthio)-2-propoxybenzaldehyde ≥97%
3-(Methylthio)-2-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂S
MW: 210.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768586
CAS Number: 2901098-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR