2-Chloro-3-methyl-5-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

81078-500MG 81078-1G 81078-5G
77336-128EA 376.98 USD
77336-128 77358-796 77382-714
2-Chloro-3-methyl-5-(methylthio)pyridine ≥97%
2-Chloro-3-methyl-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNS
MW: 173.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26127747
CAS Number: 1820706-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR