2-Isopropoxy-5-Isopropylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

81070-500MG 81070-1G 81070-5G
77338-440EA 487.19 USD
77338-440 77358-794 77382-712
2-Isopropoxy-5-Isopropylbenzoic acid ≥95%
2-Isopropoxy-5-Isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14538309
CAS Number: 1216080-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR