(4-Fluoro-5-isobutoxy-1,3-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

81064-500MG 81064-1G 81064-5G
77340-270EA 869.32 USD
77340-270 77358-792 77382-710
(4-Fluoro-5-isobutoxy-1,3-phenylene)bis(methylsulfane) ≥97%
(4-Fluoro-5-isobutoxy-1,3-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₁₂H₁₇FOS₂
MW: 260.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768577
CAS Number: 2901099-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR