4,5-Difluorophthalaldehyde ≥97%

Supplier: AOBChem USA

Warning

81051-500MG 81051-1G 81051-5G 81051-25G
77334-578EA 281.19 USD
77334-578 77358-786 77370-580 77391-266
4,5-Difluorophthalaldehyde ≥97%
4,5-Difluorophthalaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34768565
CAS Number: 854519-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR