2-Bromo-4-fluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

81029-1G 81029-5G 81029-25G
77341-442EA 85.49 USD
77341-442 77370-578 77382-704
2-Bromo-4-fluoro-N-methoxy-N-methylbenzamide ≥97%
2-Bromo-4-fluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO₂
MW: 262.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847457
CAS Number: 1178776-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR