2-Chloro-3-fluoro-N-methoxy-N-methylisonicotinamide ≥97%

Supplier: AOBChem USA

Warning

81024-500MG 81024-1G 81024-5G 81024-25G
77334-576EA 99.91 USD
77334-576 77342-902 77370-576 77382-702
2-Chloro-3-fluoro-N-methoxy-N-methylisonicotinamide ≥97%
2-Chloro-3-fluoro-N-methoxy-N-methylisonicotinamide

Precaución: for Research Use Only

Formula: C₈H₈ClFN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18257922
CAS Number: 1256790-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR