1-(Difluoromethoxy)-3-fluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

80972-500MG 80972-1G 80972-5G
77339-426EA 588.13 USD
77339-426 77358-782 77382-700
1-(Difluoromethoxy)-3-fluoro-5-iodobenzene ≥97%
1-(Difluoromethoxy)-3-fluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₇H₄F₃IO
MW: 288.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396812
CAS Number: 1261583-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR