(3-(Difluoromethoxy)-5-fluorophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

80970-500MG 80970-1G 80970-5G 80970-25G
77334-572EA 182.31 USD
77334-572 77347-046 77370-572 77391-262
(3-(Difluoromethoxy)-5-fluorophenyl)(methyl)sulfane ≥95%
(3-(Difluoromethoxy)-5-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₇F₃OS
MW: 208.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789726
CAS Number: 1804886-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR