2-Hydroxy-3-methoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

80962-500MG 80962-1G 80962-5G 80962-25G
77334-570EA 217.33 USD
77334-570 77358-780 77370-570 77391-260
2-Hydroxy-3-methoxy-5-methylbenzoic acid ≥97%
2-Hydroxy-3-methoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD11203629
CAS Number: 4386-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR