1-Bromo-2-chloro-4-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

80957-1G 80957-5G 80957-25G
77341-582EA 93.73 USD
77341-582 77370-568 77382-698
1-Bromo-2-chloro-4-propoxybenzene ≥97%
1-Bromo-2-chloro-4-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23162478
CAS Number: 1353776-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR