2-Fluoro-1-(2-fluorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

80930-1G 80930-5G 80930-25G
77341-182EA 73.13 USD
77341-182 77370-566 77382-696
2-Fluoro-1-(2-fluorophenyl)ethanone ≥97%
2-Fluoro-1-(2-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MW: 156.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24713349
CAS Number: 1402412-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR