4,5-Difluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

80773-500MG 80773-1G 80773-5G
77336-848EA 394.49 USD
77336-848 77358-778 77382-694
4,5-Difluoro-2-methylbenzaldehyde ≥97%
4,5-Difluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MW: 156.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16250159
CAS Number: 1253195-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR