5-Iodo-7-(trifluoromethyl)-1H-indazole

Supplier: AOBChem USA

Warning

80700-500MG 80700-1G 80700-5G 80700-25G
77334-564EA 222.48 USD
77334-564 77358-772 77370-560 77391-254
5-Iodo-7-(trifluoromethyl)-1H-indazole
5-Iodo-7-(trifluoromethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₄F₃IN₂
MW: 312.03 g/mol
Storage Temperature: Ambient
CAS Number: 2918965-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR