4-(Difluoromethyl)-1-iodo-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

80690-500MG 80690-1G 80690-5G
77340-380EA 978.5 USD
77340-380 77358-770 77382-692
4-(Difluoromethyl)-1-iodo-2-methylbenzene ≥95%
4-(Difluoromethyl)-1-iodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₇F₂I
MW: 268.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18395537
CAS Number: 1261645-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR