1-Bromo-3-isopropoxy-2,4-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

80679-500MG 80679-1G 80679-5G
77334-562EA 342.99 USD
77334-562 77358-764 77382-686
1-Bromo-3-isopropoxy-2,4-dimethoxybenzene ≥95%
1-Bromo-3-isopropoxy-2,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO₃
MW: 275.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27966093
CAS Number: 114605-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR