2-(Benzyloxy)-4-bromo-1,3-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

80678-500MG 80678-1G 80678-5G 80678-25G
77334-560EA 141.11 USD
77334-560 77344-938 77370-558 77391-252
2-(Benzyloxy)-4-bromo-1,3-dimethoxybenzene ≥97%
2-(Benzyloxy)-4-bromo-1,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO₃
MW: 323.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768440
CAS Number: 24534-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR