1-Bromo-3-ethoxy-2,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

80677-500MG 80677-1G 80677-5G 80677-25G
77334-558EA 186.43 USD
77334-558 77358-762 77370-556 77391-250
1-Bromo-3-ethoxy-2,4-dimethoxybenzene ≥97%
1-Bromo-3-ethoxy-2,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₃
MW: 261.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768439
CAS Number: 18111-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR