5-Bromo-3-fluoro-2-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

80646-500MG 80646-1G 80646-5G
77336-574EA 388.31 USD
77336-574 77358-760 77382-682
5-Bromo-3-fluoro-2-iodobenzoic acid ≥97%
5-Bromo-3-fluoro-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrFIO₂
Storage Temperature: Ambient
MDL Number: MFCD27943823
CAS Number: 1335511-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR