(4-(tert-Butyl)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

80629-500MG 80629-1G 80629-5G 80629-25G
77334-554EA 220.42 USD
77334-554 77358-758 77370-552 77391-248
(4-(tert-Butyl)phenyl)trimethylsilane ≥95%
(4-(tert-Butyl)phenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₃H₂₂Si
MW: 206.399 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201824
CAS Number: 18412-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR