2-Chloro-3-fluoro-4-iodo-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

80612-500MG 80612-1G 80612-5G 80612-25G
77334-550EA 99.91 USD
77334-550 77342-900 77370-548 77382-680
2-Chloro-3-fluoro-4-iodo-1-methylbenzene ≥97%
2-Chloro-3-fluoro-4-iodo-1-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFI
MW: 270.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751654
CAS Number: 909122-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR