3-Chloro-2-fluoro-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

80610-1G 80610-5G 80610-25G
77341-976EA 110.21 USD
77341-976 77370-546 77382-678
3-Chloro-2-fluoro-4-methylbenzoic acid ≥97%
3-Chloro-2-fluoro-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712064
CAS Number: 1427416-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR