1,2-Dibromo-4-chloro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

80563-500MG 80563-1G 80563-5G
77336-290EA 384.19 USD
77336-290 77358-752 77382-674
1,2-Dibromo-4-chloro-3-methylbenzene ≥97%
1,2-Dibromo-4-chloro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂Cl
Storage Temperature: Ambient
MDL Number: MFCD34768397
CAS Number: 2385526-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR