2,3-Dibromo-6-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,2-Dibromo-4-iodo-3-methylbenzene

80562-500MG 80562-1G 80562-5G 80562-25G
77334-546EA 100.94 USD
77334-546 77342-966 77370-542 77382-672
2,3-Dibromo-6-iodotoluene ≥97%
2,3-Dibromo-6-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂I
MW: 375.827 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845873
CAS Number: 1160574-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR