1,2,4-Tribromo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

80561-500MG 80561-1G 80561-25G 80561-5G
77334-544EA 301.79 USD
77334-544 77358-750 77391-242 77382-670
1,2,4-Tribromo-3-methylbenzene ≥97%
1,2,4-Tribromo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₃
Storage Temperature: Ambient
MDL Number: MFCD18400102
CAS Number: 116779-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR