1-Bromo-5-(tert-butyl)-2-chloro-3-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

80560-500MG 80560-1G 80560-5G
77334-542EA 341.96 USD
77334-542 77358-748 77382-668
1-Bromo-5-(tert-butyl)-2-chloro-3-(methylsulfonyl)benzene ≥97%
1-Bromo-5-(tert-butyl)-2-chloro-3-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrClO₂S
MW: 325.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768396
CAS Number: 2901085-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR