(2-Bromo-3-ethoxy-6-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

80489-1G 80489-5G 80489-25G
77341-490EA 89.61 USD
77341-490 77370-526 77382-652
(2-Bromo-3-ethoxy-6-fluorophenyl)trimethylsilane ≥97%
(2-Bromo-3-ethoxy-6-fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₁H₁₆BrFOSi
Storage Temperature: Ambient
MDL Number: MFCD34768351
CAS Number: 2901083-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR