(E)-4-bromobenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

80406-1G 80406-5G 80406-25G
77341-532EA 91.67 USD
77341-532 77370-516 77382-640
(E)-4-bromobenzaldehyde oxime ≥97%
(E)-4-bromobenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₇H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD00463678
CAS Number: 40979-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR