1-(3-Chloro-2,4-dimethylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

80385-500MG 80385-1G 80385-5G 80385-25G
77334-536EA 182.31 USD
77334-536 77347-044 77370-514 77391-240
1-(3-Chloro-2,4-dimethylphenyl)ethanol ≥97%
1-(3-Chloro-2,4-dimethylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO
MW: 184.663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768294
CAS Number: 2901082-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR