(3-Chloro-2,4-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

80384-500MG 80384-1G 80384-5G 80384-25G
77334-534EA 230.72 USD
77334-534 77358-746 77370-512 77391-238
(3-Chloro-2,4-dimethylphenyl)(methyl)sulfane ≥97%
(3-Chloro-2,4-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClS
MW: 186.702 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489463
CAS Number: 1822987-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR