2-Chloro-4-iodo-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

80383-500MG 80383-1G 80383-5G 80383-25G
77334-532EA 189.52 USD
77334-532 77358-744 77370-510 77391-236
2-Chloro-4-iodo-1,3-dimethylbenzene ≥97%
2-Chloro-4-iodo-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClI
Storage Temperature: Ambient
MDL Number: MFCD34768293
CAS Number: 1222817-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR