(2-Chloro-4-fluoro-6-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

80366-500MG 80366-1G 80366-5G 80366-25G
77334-530EA 281.19 USD
77334-530 77358-742 77370-506 77391-234
(2-Chloro-4-fluoro-6-methylphenyl)(methyl)sulfane ≥95%
(2-Chloro-4-fluoro-6-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFS
MW: 190.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752227
CAS Number: 1807268-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR