tert-Butyl (2,4-dimethylphenyl)carbamate ≥97%

Supplier: AOBChem USA

Warning

80359-500MG 80359-1G 80359-5G 80359-25G
77334-528EA 133.9 USD
77334-528 77344-422 77370-504 77391-232
tert-Butyl (2,4-dimethylphenyl)carbamate ≥97%
tert-Butyl (2,4-dimethylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD07127704
CAS Number: 129822-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR