4-Fluoro-5-iodo-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

80331-500MG 80331-1G 80331-5G
77339-346EA 579.89 USD
77339-346 77358-740 77382-636
4-Fluoro-5-iodo-2-methoxybenzaldehyde ≥97%
4-Fluoro-5-iodo-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29922368
CAS Number: 1956319-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR