2-Fluoro-5-formyl-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

80330-500MG 80330-1G 80330-5G
77339-540EA 595.34 USD
77339-540 77358-738 77382-634
2-Fluoro-5-formyl-4-methoxybenzoic acid ≥97%
2-Fluoro-5-formyl-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₄
MW: 198.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34768283
CAS Number: 2386444-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR