N-(2-cyanophenyl)benzamide ≥95%

Supplier: AOBChem USA

Warning

80321-500MG 80321-1G 80321-5G 80321-25G
77334-524EA 120.51 USD
77334-524 77343-840 77370-500 77382-630
N-(2-cyanophenyl)benzamide ≥95%
N-(2-cyanophenyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₂O
MW: 222.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00451472
CAS Number: 40288-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR