3'-Bromo-2,3-dimethoxy-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

80311-500MG 80311-1G 80311-5G
77337-606EA 426.42 USD
77337-606 77358-732 77382-626
3'-Bromo-2,3-dimethoxy-1,1'-biphenyl ≥95%
3'-Bromo-2,3-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
MW: 293.156 g/mol
Storage Temperature: Ambient
CAS Number: 2138009-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR