7-Bromo-4-chloro-1H-indazol-3-amine ≥97%

Supplier: AOBChem USA

Warning

80287-500MG 80287-1G 80287-5G
77338-678EA 507.79 USD
77338-678 77358-726 77382-620
7-Bromo-4-chloro-1H-indazol-3-amine ≥97%
7-Bromo-4-chloro-1H-indazol-3-amine

Precaución: for Research Use Only

Formula: C₇H₅BrClN₃
MW: 246.492 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32705599
CAS Number: 1626336-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR