7-Bromo-5-methyl-1H-indazol-3-amine ≥97%

Supplier: AOBChem USA

Warning

80286-500MG 80286-1G 80286-5G
77339-992EA 704.52 USD
77339-992 77358-724 77382-618
7-Bromo-5-methyl-1H-indazol-3-amine ≥97%
7-Bromo-5-methyl-1H-indazol-3-amine

Precaución: for Research Use Only

Formula: C₈H₈BrN₃
Storage Temperature: Ambient
MDL Number: MFCD22565599
CAS Number: 1388073-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR