2-(Phenylethynyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

80284-1G 80284-5G 80284-25G
77341-722EA 101.97 USD
77341-722 77370-492 77382-616
2-(Phenylethynyl)benzonitrile ≥95%
2-(Phenylethynyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₅H₉N
Storage Temperature: Ambient
MDL Number: MFCD01319642
CAS Number: 32183-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR