tert-Butyl (2,4-difluorophenyl)carbamate ≥95%

Supplier: AOBChem USA

Warning

80248-500MG 80248-1G 80248-5G
77337-030EA 405.82 USD
77337-030 77358-720 77382-610
tert-Butyl (2,4-difluorophenyl)carbamate ≥95%
tert-Butyl (2,4-difluorophenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD21170345
CAS Number: 701269-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR