1-Bromo-3-ethyl-2-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

79974-500MG 79974-1G 79974-5G
77334-514EA 313.12 USD
77334-514 77358-714 77382-606
1-Bromo-3-ethyl-2-fluoro-5-methylbenzene ≥97%
1-Bromo-3-ethyl-2-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD34768231
CAS Number: 2705745-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR