tert-Butyl (2-bromo-4-chlorophenyl)carbamate ≥95%

Supplier: AOBChem USA

Warning

79969-500MG 79969-1G 79969-5G 79969-25G
77334-512EA 174.07 USD
77334-512 77346-464 77370-488 77391-220
tert-Butyl (2-bromo-4-chlorophenyl)carbamate ≥95%
tert-Butyl (2-bromo-4-chlorophenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD11975574
CAS Number: 384793-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR