(E)-1-(2-bromophenyl)-3-(dimethylamino)prop-2-en-1-one ≥97%

Supplier: AOBChem USA

Warning

79956-500MG 79956-1G 79956-5G 79956-25G
77334-510EA 220.42 USD
77334-510 77358-710 77370-486 77391-218
(E)-1-(2-bromophenyl)-3-(dimethylamino)prop-2-en-1-one ≥97%
(E)-1-(2-bromophenyl)-3-(dimethylamino)prop-2-en-1-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO
MW: 254.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998336
CAS Number: 1203608-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR