1-Bromo-2-chloro-3-(difluoromethoxy)-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

79946-500MG 79946-1G 79946-5G
77334-508EA 312.09 USD
77334-508 77358-708 77382-602
1-Bromo-2-chloro-3-(difluoromethoxy)-4-fluorobenzene ≥97%
1-Bromo-2-chloro-3-(difluoromethoxy)-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD34768223
CAS Number: 2386010-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR