1,3-Dibromo-2-fluoro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

79915-500MG 79915-1G 79915-5G 79915-25G
77334-504EA 234.84 USD
77334-504 77358-706 77370-480 77391-214
1,3-Dibromo-2-fluoro-4-methylbenzene ≥97%
1,3-Dibromo-2-fluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
MW: 267.921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19686160
CAS Number: 1356114-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR