3-Bromo-2-fluoro-4-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

79910-500MG 79910-1G 79910-5G 79910-25G
77334-502EA 181.28 USD
77334-502 77346-876 77370-476 77391-212
3-Bromo-2-fluoro-4-methylbenzoic acid ≥95%
3-Bromo-2-fluoro-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712066
CAS Number: 1427382-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR