5-Iodo-2-(4-methylpiperazin-1-yl)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

79856-500MG 79856-1G 79856-25G 79856-5G
77334-498EA 307.97 USD
77334-498 77358-704 77391-208 77382-596
5-Iodo-2-(4-methylpiperazin-1-yl)pyrimidine ≥97%
5-Iodo-2-(4-methylpiperazin-1-yl)pyrimidine

Precaución: for Research Use Only

Formula: C₉H₁₃IN₄
MW: 304.131 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870536
CAS Number: 1379295-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR