Methyl 4-formyl-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

79852-500MG 79852-1G 79852-25G 79852-5G
77334-496EA 300.76 USD
77334-496 77358-702 77391-206 77382-594
Methyl 4-formyl-3-(trifluoromethyl)benzoate ≥97%
Methyl 4-formyl-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O₃
MW: 232.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381355
CAS Number: 1823806-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR